Neutralisers for alkaline detergents
148-1843
- Description:Neutraliser, Sekumatic® FNZ, for surgical instruments
- Packed:5 l can
- Pk:5.000 mL
Ideal for surgical instruments, MIC, rigid endoscopes, plastic utensils and elastomers (rubber) for anesthesia, surgery, urology, surgical shoes, baby bottles and laboratory glassware.
- Citric acid based
- High environmental compatibility
- Surfactant
- Concentration: 1 to 3 ml/L
- Suitable for all water hardnesses