Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Stains
Microscopy and staining reagents
Mounting media
Mounting media
Print
Mounting media
Fluoro-Gel II mounting media, 100 mL
Catalog # SCIAE17985-51
Supplier:
SCI MUENCHEN
Mounting media
Catalog # SCIAE17985-51
Supplier:
SCI MUENCHEN
CAS Number:
Expand image
Return to Top
Mounting media
Fluoro-Gel II mounting media, 100 mL
Specifications
Add to cart
Jump to:
Specifications
CAS Number:
Price
Inquire For Price
Log in
for availability
Unit of Measure
1 * 100 mL
-
+
Inquire For Price
Return Policy
Product Details & Documents
Specifications
Specifications
Pk:
100 mL
Description:
Fluoro-Gel II mounting media
Specifications