Test kits, arsenic
Catalog # LOVI400700
Supplier: Lovibond
CAS Number:
Specifications
- Pk:100 Tests
- Description:Arsenic test kit
- Range:0,0,005, 0,01, 0,025, 0,05, 0,1, 0,25, 0,5 mg/l, As3+/5+
Specifications
About this item
High sensitivity kit suitable for the determination of arsenic in drinking water.
- Detects 0,005 mg/l arsenic
- A solid acid substance is used in order to avoid any irritation by a corrosive acid on the user’s hands
- The unbreakable plastic reaction vessel is more convenient and safe for on-site testing
Delivery: The test kit contains a water-proof colour chart which also includes the brief instruction for use in pictograms. Supplied in a carrying case.