
3,4-Ethylenedioxythiophene 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:126213-50-1
:
Formula:C₆H₆O₂S
MDL Number:MFCD02093622
:142.17 g/mol
Boiling Pt:193 °C
Melting Pt:10.5…12 °C
Density:1.331 g/cm³
Flash Pt:109 °C
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:Colorless to yellow Liquid
- Infrared spectrum:Conforms
- GC:≥98.5 %