Order Entry
Export
Orders LinkContactUsLinkComponent
 

5,7-Dichloropyrazolo[1,5-a]pyrimidine 95%

Glass bottle, 1 g
Catalog # ACRO454130010
Irritant
5,7-Dichloropyrazolo[1,5-a]pyrimidine 95%
Catalog # ACRO454130010
CAS Number:  57489-77-7
5,7-Dichloropyrazolo[1,5-a]pyrimidine 95%
5,7-Dichloropyrazolo[1,5-a]pyrimidine 95%

Glass bottle, 1 g

CAS Number:  57489-77-7
Price
406,00 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:57489-77-7
  • Formula:C₆H₃Cl₂N₃
  • MDL Number:MFCD08273918
  • Molecular Weight:188.0141 g/mol
  • Melting Pt:75…76.5 °C
  • Storage Temperature:Ambient

 

Specifications

Specification Test Results

  • Appearance:
    White to yellow Solid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥94.0 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g

Specifications