Order Entry
Export
ContactUsLinkComponent
 
 
2,5-Dichloropyrimidine 95%
  ACRO454040010
 :  
undefined
 
2,5-Dichloropyrimidine 95%
  ACRO454040010
 :  

 

  • Pk:
    1 g
  • Seq:
    3047

 

 

  • Appearance:
    White to yellow Powder or low melting solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    49 to 56 °C
  • GC:
    ≥94.0 %

 

 

 :
 :148.97 g/mol
 :52…55 °C
 :GHS07
 :Ambient
 :22536-67-0
 :C₄H₂Cl₂N₂
 :MFCD00673131