Order Entry
Export
Orders LinkContactUsLinkComponent
 

Lead(II) sulphide 90%, pure

Irritant
Health
Environment
Lead(II) sulphide 90%, pure
Certificates
Lead(II) sulphide 90%, pure

2 Options Available Below

Product Details & Documents
Synonyms:  Lead sulphide
  • CAS Number:1314-87-0
  • Formula:̍PbS
  • MDL Number:MFCD00016280
  • Molecular Weight:239.27 g/mol
  • Boiling Pt:1281.1 °C
  • Melting Pt:1113.9 °C
  • Density:7,5 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • UN:3077
  • ADR:9,III

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    Grey to purple Powder
  • Assay:
    ≥78 % (Pb)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 99,40 €
    99,40 €
    1 * 1 kg
       99,40 €

  • Plastic bottle
    5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 462,00 €
    462,00 €
    1 * 5 kg
       462,00 €