
2,4-Dibromopyridine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:58530-53-3
:
Formula:C₅H₃Br₂N
MDL Number:MFCD01859720
Melting Pt:36…38.5 °C
UN:2811
ADR:6.1,III
:236.89 g/mol
:GHS06
Storage Temperature:Ambient
Specification Test Results
- Appearance:Off-white to yellow to brown Low melting solid
- Infrared spectrum:Conforms
- Melting point:35 to 40 °C
- GC:≥96.0 %