
(4,4-Difluorocyclohexyl)methanol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:178312-48-6
:
Formula:C₇H₁₂F₂O
MDL Number:MFCD11847775
:150.17 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to yellow Viscous liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %