
2,2-Difluoro-1,3-benzodioxole-5-carboxylic acid 97%
Catalog # ACRO449140050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Light grey to tan Powder
- Infrared spectrum:Conforms
- Melting point:154 to 157 °C
- HPLC:≥96.0 %
Specification Test Results
About this item
:
:202.12 g/mol
Melting Pt:153…157 °C
:GHS07
Storage Temperature:Ambient
CAS Number:656-46-2
Formula:C₈H₄F₂O₄
MDL Number:MFCD00792417