

2,6-Dimethoxyaniline 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2734-70-5
:
Formula:C₈H₁₁NO₂
MDL Number:MFCD00053934
:153.18 g/mol
Melting Pt:76…78 °C
UN:2811
ADR:6.1,III
:GHS06
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Grey to brown Crystalline powder
- Infrared spectrum:Conforms
- GC:≥95 %