
3,3',5,5'-Tetra-tert-butyl-4,4'-diphenoquinone 98%
ACRO448300020
:
- Pack type:Glass bottle
- Pk:2 g
- Seq:3107
- Appearance:Yellow to brown to red Powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
:
:247…248 °C
:408.63 g/mol
:GHS07
:Ambient
:2455-14-3
:C₂₈H₄₀O₂
:MFCD00051798