Sodium sulphite 98% for biochemistry
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7757-83-7
:
Formula:Na₂SO₃
MDL Number:MFCD00003503
:126.04 g/mol
Melting Pt:>500 °C (dec.)
Density:2.63 g/cm³
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder or crystals
- Titration Iodimetric:≥97.5 %
- Solubility:(1 M in water) Clear colorless
- Chloride (Cl):≤0.02 %
- Arsenic (As):≤1 ppm
- Copper (Cu):≤5 ppm
- Iron (Fe):≤10 ppm
- Lead (Pb):≤10 ppm
- Selenium (Se):≤10 ppm
- Zinc (Zn):≤10 ppm
- Heavy metals (as Pb):≤10 ppm