Order Entry
Export
ContactUsLinkComponent
4,4-Dimethylcyclohex-2-en-1-on-2-ylboronic acid pinacol ester 97%
Irritant
4,4-Dimethylcyclohex-2-en-1-on-2-ylboronic acid pinacol ester 97%
Catalog # ACRO444980010
CAS Number:  
undefined
Irritant
4,4-Dimethylcyclohex-2-en-1-on-2-ylboronic acid pinacol ester 97%
Catalog # ACRO444980010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to yellow Solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    98 to 102 °C
  • GC:
    ≥96.0 %
  • NMR (1H-NMR):
    Consistent with structure

Specification Test Results

About this item

 :
 :250.1400 g/mol
Melting Pt:99…102 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:219489-09-5
Formula:C₁₄H₂₃BO₃
MDL Number:MFCD09027074