Order Entry
Export
ContactUsLinkComponent
Petroleum spirit, 100…140 °C, Technical Grade
Irritant
Health
Environment
Flammable
Petroleum spirit, 100…140 °C, Technical Grade
Catalog # ACRO444360250
CAS Number:  
undefined
Irritant
Health
Environment
Flammable
Petroleum spirit, 100…140 °C, Technical Grade
Catalog # ACRO444360250
CAS Number:  

Specifications

  • Pack type:
    Metal drum
  • Pk:
    25 L
  • Seq:
    5327

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Refractive index:
    1.4070 to 1.4150 (20 °C, 589 nm)
  • Boiling point:
    98 to 140 °C (760 mmHg)
  • Density:
    0.737 to 0.747 kg/l

Specification Test Results

About this item

 :
Boiling Pt:100…140 °C (760 mmHg)
Melting Pt:–30 °C
Density:0.74 g/cm³
Flash Pt:2 °C
Storage Temperature:Ambient
UN:3295
ADR:3,II
 :GHS07
 :GHS08
 :GHS09
 :GHS02
CAS Number:64742-49-0
 :8032-32-4
MDL Number:MFCD00081849