Order Entry
Export
ContactUsLinkComponent
 
Irritant
Paraffin, liquid, Technical Grade
Catalog # ACRO444180250
CAS Number:  
undefined
Irritant
Paraffin, liquid, Technical Grade
Catalog # ACRO444180250
CAS Number:  

Specifications

  • Pack type:
    Plastic can
  • Pk:
    25 L
  • Seq:
    5327

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Oily liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4600 to 1.4800 (20°C, 589 nm)
  • Density:
    (20°C) 0.810 to 0.890 g/ml
  • Viscosity:
    20 to 80 mm²/s (40°C)

Specification Test Results

About this item

 :
Boiling Pt:300 °C (760 mmHg)
Melting Pt:–24 °C
Density:0.88 g/cm³
Flash Pt:218 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:8012-95-1
 :8002-74-2