
3-(N-Boc-aminomethyl)-5-bromopyridine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:943722-24-5
:
Formula:C₁₁H₁₅BrN₂O₂
MDL Number:MFCD10700149
:287.157 g/mol
Melting Pt:84.5…86 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to yellow Powder
- Infrared spectrum:Conforms
- GC:≥96.0 %