
5-Bromo-2,4-dimethoxypyrimidine 97%
Catalog # ACRO443390050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Melting point:62 to 67 °C
- GC:≥96.0 %
Specification Test Results
About this item
:
:219.0400 g/mol
Boiling Pt:125 °C (17Torr)
Melting Pt:63…65 °C
:GHS07
Storage Temperature:Ambient
CAS Number:56686-16-9
Formula:C₆H₇BrN₂O₂
MDL Number:MFCD00038016