Order Entry
Export
ContactUsLinkComponent
 
 
2-Chloro-4,6-dimethylpyrimidine 97%
 
undefined
 
2-Chloro-4,6-dimethylpyrimidine 97%

 

  • Appearance:
    White to yellow Crystalline powder or low melting solid
  • Infrared spectrum:
    Conforms
  • Melting point:
    34 to 39 °C
  • GC:
    ≥96.0 %