
6-Nitro-2,4-xylidine 97%
ACRO442440050
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:Light yellow to orange Solid
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
:
:166.18 g/mol
:69.5…70.5 °C
:Ambient
:2811
:6.1,III
:GHS07
:1635-84-3
:C₈H₁₀N₂O₂
:MFCD00007739