
4-Bromo-2-methoxyphenol 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7368-78-7
:
Formula:C₇H₇BrO₂
MDL Number:MFCD00051937
:203.04 g/mol
Boiling Pt:90…92 °C (1hPa)
Melting Pt:35…39 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to white to light brown Low melting crystalline solid
- Infrared spectrum:Conforms
- HPLC:≥97.5 %