
(3,4-Dimethoxybenzyl)methylamine 97%
Catalog # ACRO439680050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to yellow to light brown Liquid
- Infrared spectrum:Conforms
- GC:≥96.0 %
Specification Test Results
About this item
:
:181.24 g/mol
Flash Pt:110 °C
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
CAS Number:63-64-9
Formula:C₁₀H₁₅NO₂
MDL Number:MFCD00837593