1,1'-Oxalyldiimidazole, Technical Grade
Catalog # ACRO439620050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Yellow to brown Powder
- Infrared spectrum:Conforms
- HPLC:≥80.0 %
Specification Test Results
About this item
:
:190.16 g/mol
Melting Pt:114…117 °C
Storage Temperature:Ambient
UN:3263
ADR:8,III
:GHS05
:GHS08
CAS Number:18637-83-7
Formula:C₈H₆N₄O₂
MDL Number:MFCD00043067