
7-Chloropyrazolo[1,5-a]pyrimidine 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:58347-49-2
:
Formula:C₆H₄ClN₃
MDL Number:MFCD04035684
Melting Pt:98…102 °C
UN:3261
ADR:8,III
:153.5700 g/mol
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to yellow Solid (May have a pink hue)
- Infrared spectrum:Conforms
- Melting point:99 to 103 °C
- GC:≥94.0 %