Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl picolinate
Methyl picolinate 98%
Methyl picolinate 98%
:
Thermo Fisher Scientific
Methyl picolinate 98%
:
Thermo Fisher Scientific
:
2459-07-6
:
:
C₇H₇NO₂
:
MFCD00038038
:
137.14 g/mol
:
95 °C (1hPa)
:
14…16 °C
:
1.137 g/cm³
:
103 °C
:
GHS07
:
Ambient
Appearance:
Clear colorless to yellow to orange Liquid
Infrared spectrum:
Conforms
GC:
≥97.5 %
ACRO43814