Order Entry
Export
ContactUsLinkComponent
 
Irritant
4-Chloro-2,5-dimethoxyaniline 97%
Catalog # ACRO437861000
CAS Number:  
undefined
Irritant
4-Chloro-2,5-dimethoxyaniline 97%
Catalog # ACRO437861000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Appearance:
    Off-white to brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    118 to 120 °C
  • HPLC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :187.63 g/mol
Melting Pt:118…120 °C
Flash Pt:163 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:6358-64-1
Formula:C₈H₁₀ClNO₂
MDL Number:MFCD00014893