Order Entry
Export
ContactUsLinkComponent
 
 
2,6-Pyridinedicarboxaldehyde 97%
 
undefined
 
2,6-Pyridinedicarboxaldehyde 97%

 

  • Appearance:
    White to tan Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    122 to 126 °C
  • GC:
    ≥96.0 %