Iron 99.99% (trace metals basis), granules
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7439-89-6
:
Formula:Fe
MDL Number:MFCD00010999
:55.85 g/mol
Boiling Pt:3000 °C (760 mmHg)
Melting Pt:1535 °C
Density:7,86 g/cm³ (20 °C)
Storage Temperature:Ambient
Specification Test Results
- Appearance:Silver grey Granules
- Particle size:0.5 to 1 mm
- Total trace metal impurities:≤100 ppm