


Potassium tetrachloroplatinate(II) 99.99% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10025-99-7
:
Formula:Cl₄K₂Pt
MDL Number:MFCD00011378
:415.09 g/mol
Boiling Pt:35 - 40 °C (20 -rr)
Melting Pt:250 °C (dec.)
Density:3,38 g/cm³ (20 °C)
UN:3288
ADR:6.1,III
:GHS06
:GHS08
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:Orange to red to brown Crystals
- Total trace metal impurities:≤100 ppm