Order Entry
Export
ContactUsLinkComponent
Glycerol-1,2-carbonate 90%
Glycerol-1,2-carbonate 90%
Catalog # ACRO436170250
CAS Number:  
undefined
Glycerol-1,2-carbonate 90%
Catalog # ACRO436170250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Elemental Analysis:
    36.6 to 44.7 % (C)

Specification Test Results

About this item

 :
 :118.09 g/mol
Boiling Pt:142.5 °C (1 mmHg)
Density:1.4 g/cm³
Flash Pt:110 °C
Storage Temperature:Ambient
CAS Number:931-40-8
Formula:C₄H₆O₄
MDL Number:MFCD00085561