Order Entry
Export
Orders LinkContactUsLinkComponent
Estradiol 98%

Estradiol 98%

Health
Estradiol 98%
Certificates
Estradiol 98%

2 Options Available Below

Product Details & Documents
Synonyms:  beta-Estradiol, 17β-Estradiol
  • CAS Number:50-28-2
  • Formula:C₁₈H₂₄O₂
  • MDL Number:MFCD00003693
  • Molecular Weight:272.39 g/mol
  • Melting Pt:176…180 °C
  • Storage Temperature:Ambient

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    173 to 179 °C
  • Water:
    ≤3.5 %
  • HPLC:
    ≥97.5 %
  • Specific optical rotation:
    +76° to +83° (20 °C, 589 nm) (c=1, dioxane)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 172,00 €
    172,00 €
    1 * 5 g
       172,00 €

  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 628,00 €
    628,00 €
    1 * 25 g
       628,00 €