



3,4-Dimethylphenylmagnesium bromide 0.5 M in THF, AcroSeal®
Catalog # 431910500.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:50 mL
- Seq:2370
Specifications
Specification Test Results
- Appearance:Yellow to brown Solution
- Titration Grignard:0.40 to 0.60 M
- Refractive index:1.4590 to 1.4610 (20°C, 589 nm)
- GC:≥98.0 %
Specification Test Results
About this item
:
:209.369 g/mol
Boiling Pt:65 °C (760 mmHg)
Density:0.91 g/cm³
Flash Pt:–17 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
:GHS07
:GHS05
:GHS02
:GHS08
CAS Number:89980-68-7
Formula:C₈H₉BrMg
MDL Number:MFCD01311496