


Hydrobromic acid 33% (w/w) in glacial acetic acid, AcroSeal®, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10035-10-6
Formula:HBr
MDL Number:MFCD00011323
:80.91 g/mol
Density:1.42 g/cm³
Flash Pt:39 °C
UN:2920
ADR:8,II
:GHS07
:GHS05
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear yellow to yellow-orange Solution (May darken on storage)
- Water:≤0.5 %
- Titration with NaOH:31.5 to 36.5 %
- HPLC:≥97.0 %