
5-Amino-1,4-benzodioxane 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16081-45-1
:
Formula:C₈H₉NO₂
MDL Number:MFCD03695459
ADR:6.1,III
:151.1626 g/mol
UN:2810
:GHS06
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear brown to purple Liquid
- Infrared spectrum:Conforms
- GC:≥94.0 %
- Sulfite (SO3):≤5 ppm
- Sulfate (SO4):≤5 ppm
- Arsenic (As):≤0.5 ppm
- Iron (Fe):≤10 ppm
- Heavy metals (as Pb):≤5 ppm