Chlorotrimethylsilane 98%, AcroSeal®
Catalog # 426431000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3870 to 1.3890 (20°C, 589 nm)
- GC:≥97.5 %
Specification Test Results
About this item
CAS Number:75-77-4
:
Formula:C₃H₉ClSi
MDL Number:MFCD00000502
Boiling Pt:57 °C (760 mmHg)
Melting Pt:–58 °C
Density:0.85 g/cm³
Flash Pt:–28 °C
UN:1298
ADR:3,II
:108.64 g/mol
:GHS06
:GHS05
:GHS02
Storage Temperature:Ambient