
(5R,6S)-(-)-4-Benzyloxycarbonyl-5,6-diphenyl-2-morpholinone 98%
ACRO398660010
:
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Specific optical rotation:-64° to -68° (25°C, 589 nm) (c=5.5,CH2Cl2)
:
:387.435 g/mol
:205…207 °C
:GHS07
:Refrigerator
:100516-54-9
:C₂₄H₂₁NO₄
:MFCD00074958