
3H-Imidazo[4,5-c]pyridine 97%
Catalog # ACRO398500010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Beige Powder
- Infrared spectrum:Conforms
- Melting point:168 to 172 °C
- HPLC:≥96.0 %
Specification Test Results
About this item
:
Melting Pt:168…172 °C
:119.13 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:272-97-9
Formula:C₆H₅N₃
MDL Number:MFCD00051808