Order Entry
Export
Orders LinkContactUsLinkComponent
 

2-Nitroethanol 97%

Irritant
2-Nitroethanol 97%
Certificates
2-Nitroethanol 97%

3 Options Available Below

Product Details & Documents
  • CAS Number:625-48-9
  • Formula:C₂H₅NO₃
  • MDL Number:MFCD00007405
  • Molecular Weight:91.07 g/mol
  • Boiling Pt:203…206 °C (760 mmHg)
  • Melting Pt:–80 °C
  • Density:1.292 g/cm³
  • Flash Pt:>112 °C
  • Storage Temperature:Ambient

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless to yellow to orange Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4430 to 1.4450 (20°C, 589 nm)
  • GC:
    ≥96.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 49,90 €
    49,90 €
    1 * 5 g
       49,90 €

  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 166,00 €
    166,00 €
    1 * 25 g
       166,00 €

  • Glass bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 546,00 €
    546,00 €
    1 * 100 g
       546,00 €