Order Entry
Export
ContactUsLinkComponent
(R)-(+)-3-Amino-1,2-propanediol 98%
Corrosive
(R)-(+)-3-Amino-1,2-propanediol 98%
Certificates
undefined
Corrosive
(R)-(+)-3-Amino-1,2-propanediol 98%

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    53 to 57 °C
  • Specific optical rotation:
    +27° to +29° (20 °C, 589 nm) (C=4, 5 N HCl)
  • Titration with HClO4:
    ≥97.5 %

Specification Test Results

Related Information