



Petroleum spirit, 40…60 °C, extra dry (max. 50 ppm H₂O), AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:64742-49-0
:64742-82-1
MDL Number:MFCD00081849
Boiling Pt:40…65 °C (760 mmHg)
Density:0.65 g/cm³
Flash Pt:–45 °C
Storage Temperature:Ambient
UN:1268
ADR:3,II
:GHS08
:GHS09
:GHS02
:GHS07
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.005 % (Coulometric)
- Residue after evaporation:≤0.001 %
- Filtered to 0.2 micrometer:Filtered