


(±)-2-Methyltetrahydrofuran, extra dry 99+% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:96-47-9
:
Formula:C₅H₁₀O
MDL Number:MFCD00005367
:86.13 g/mol
Boiling Pt:78…80 °C (760 mmHg)
Melting Pt:–136 °C
Density:0.86 g/cm³
Flash Pt:–11 °C
UN:2536
ADR:3,II
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.002 %
- GC:≥99.0 %
- Stabilizer:150 to 400 ppm (BHT)
- Peroxides:≤50 ppm