Order Entry
Export
ContactUsLinkComponent
1,3-Difluoro-2-nitrobenzene 98%
Irritant
1,3-Difluoro-2-nitrobenzene 98%
Catalog # ACRO397340050
CAS Number:  
undefined
Irritant
1,3-Difluoro-2-nitrobenzene 98%
Catalog # ACRO397340050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Clear yellow to orange Liquid or low melting solid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥97.5 %
  • NMR (1H-NMR):
    Consistent with structure

Specification Test Results

About this item

 :
 :159.09 g/mol
Boiling Pt:91…92 °C (11 mmHg)
Density:1.503 g/cm³
Flash Pt:88 °C
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
 :GHS07
CAS Number:19064-24-5
Formula:C₆H₃F₂NO₂
MDL Number:MFCD00192035