

N,N-Dimethylacetamide, extra dry 99.5%, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:127-19-5
:
Formula:CH₃CON(CH₃)₂
MDL Number:MFCD00008686
:87.12 g/mol
Boiling Pt:164…166 °C (760 mmHg)
Melting Pt:–20 °C
Density:0.937 g/cm³
Flash Pt:70 °C
Storage Temperature:Ambient
ADR:6.1,III
:GHS07
:GHS08
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥99.4 %
- Residue after evaporation:≤0.005 %