
3,5-Dibromobenzaldehyde 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56990-02-4
:
Formula:C₇H₄Br₂O
MDL Number:MFCD00156887
:263.92 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to light beige Powder or crystals
- Infrared spectrum:Conforms
- Melting point:88 to 92 °C
- GC:≥98.0 %