Order Entry
Export
ContactUsLinkComponent
3,7-Dimethyl-1-octanol 95%
Irritant
Environment
3,7-Dimethyl-1-octanol 95%
Certificates
undefined
Irritant
Environment
3,7-Dimethyl-1-octanol 95%

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4340 to 1.4360 (20°C, 589 nm)
  • GC:
    ≥94 %

Specification Test Results

Related Information