Order Entry
Export
Orders LinkContactUsLinkComponent
4,4'-Biphenyldicarboxaldehyde 97%

4,4'-Biphenyldicarboxaldehyde 97%

Irritant
4,4'-Biphenyldicarboxaldehyde 97%
Certificates
4,4'-Biphenyldicarboxaldehyde 97%

2 Options Available Below

Product Details & Documents
Synonyms:  [1,1'-Biphenyl]-4,4'-dicarbaldehyde, 4,4'-Diformylbiphenyl
  • CAS Number:66-98-8
  • Formula:C₁₄H₁₀O₂
  • MDL Number:MFCD00016714
  • Molecular Weight:210.23 g/mol
  • Melting Pt:144…147 °C
  • Storage Temperature:Ambient

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    144 to 147 °C
  • GC:
    ≥96.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 83,70 €
    83,70 €
    1 * 1 g
       83,70 €

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 253,00 €
    253,00 €
    1 * 5 g
       253,00 €