1,1-Dimethylurea 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:598-94-7
:
Formula:C₃H₈N₂O
MDL Number:MFCD00007959
:88.11 g/mol
Melting Pt:179…183 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:179 to 183 °C
- Titration with HClO4:≥98.0 %