



Cyclohexylmagnesium chloride 1.3 M in THF/toluene, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:931-51-1
:
Formula:C₆H₁₁ClMg
MDL Number:MFCD00003816
:142.91 g/mol
Density:0.95 g/cm³
Flash Pt:–21 °C
UN:2924
ADR:3,II
:GHS07
:GHS08
:GHS05
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear dark brown Solution (May develop some turbidity or precipitate)
- Titration Grignard:1.26 to 1.40 M