Chitosan, M.W. 100,000-300,000
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9012-76-4
:MFCD00149018
MDL Number:MFCD00161512
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Off-white to yellow to beige Powder
- Infrared spectrum:Conforms
- Loss on drying:≤10 % (2 g, 105°C, 2 h)
- Solubility:(10mg/ml, 1M acetic acid) Yellow light hazy to hazy
- Ash:≤2 %
- Degree of Deacetylation:≥75 %
- Molecular number:100000 to 300000 Da