Order Entry
Export
ContactUsLinkComponent
Bis(trimethylsilyl)malonate 98%
Flammable
Bis(trimethylsilyl)malonate 98%
Certificates
undefined
Flammable
Bis(trimethylsilyl)malonate 98%

Specification Test Results

  • Appearance:
    Clear colorless to straw yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4150 to 1.4170 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

Related Information